missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-Toluate 98.0+%, TCI America™

$29.83 - $218.10

Chemical Identifiers

CAS 99-75-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008441
InChI Key QSSJZLPUHJDYKF-UHFFFAOYSA-N
Synonym methyl p-toluate, methyl-p-toluate, p-carbomethoxytoluene, methyl 4-toluate, 4-methylbenzoic acid methyl ester, methyl p-methylbenzoate, benzoic acid, 4-methyl-, methyl ester, 4-methoxycarbonyl toluene, p-toluic acid, methyl ester, methyl p-toluenecarboxylate
PubChem CID 7455
IUPAC Name methyl 4-methylbenzoate
SMILES CC1=CC=C(C=C1)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T029725G
View Documents
TCI America
T029725G
25 g N/A
Only null left
 
T0297500G
View Documents
TCI America
T0297500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

99-75-2
150.177
QSSJZLPUHJDYKF-UHFFFAOYSA-N
7455
CC1=CC=C(C=C1)C(=O)OC
C9H10O2
MFCD00008441
methyl p-toluate, methyl-p-toluate, p-carbomethoxytoluene, methyl 4-toluate, 4-methylbenzoic acid methyl ester, methyl p-methylbenzoate, benzoic acid, 4-methyl-, methyl ester, 4-methoxycarbonyl toluene, p-toluic acid, methyl ester, methyl p-toluenecarboxylate
methyl 4-methylbenzoate

Specifications

99-75-2
223°C
MFCD00008441
methyl p-toluate, methyl-p-toluate, p-carbomethoxytoluene, methyl 4-toluate, 4-methylbenzoic acid methyl ester, methyl p-methylbenzoate, benzoic acid, 4-methyl-, methyl ester, 4-methoxycarbonyl toluene, p-toluic acid, methyl ester, methyl p-toluenecarboxylate
CC1=CC=C(C=C1)C(=O)OC
150.177
150.18
Crystals
White
C9H10O2
25 g
QSSJZLPUHJDYKF-UHFFFAOYSA-N
methyl 4-methylbenzoate
7455
≥98.0% (GC)
Methyl p-Toluate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1335&(3)-1380

RTECSNumber : XU2150000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'