missing translation for 'onlineSavingsMsg'
Learn More

Methyl-p -hydroxybenzoate, 99.83%, MP Biomedicals™

Catalog No. ICN10234101
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10234101 1 kg
ICN10234105 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10234101 Supplier MP Biomedicals Inc Supplier No. 0210234101

Chemical Identifiers

CAS 99-76-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
InChI Key LXCFILQKKLGQFO-UHFFFAOYSA-N
Synonym methylparaben, methyl paraben, methyl p-hydroxybenzoate, nipagin, methyl parahydroxybenzoate, maseptol, p-hydroxybenzoic acid methyl ester, 4-hydroxybenzoic acid methyl ester, p-carbomethoxyphenol, tegosept m
PubChem CID 7456
ChEBI CHEBI:31835
IUPAC Name methyl 4-hydroxybenzoate
SMILES COC(=O)C1=CC=C(C=C1)O

Specifications

Color White
Quantity 1 kg
Formula Weight 152.1
Percent Purity 99.83%
Physical Form Crystalline Powder
Chemical Name or Material Methyl-p-hydroxybenzoate