missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-Anisate 99.0+%, TCI America™

Catalog No. A1125500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1125500G 500 g
A112525G 25 g
A1125100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A1125500G Supplier TCI America Supplier No. A1125500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-98-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00008437
InChI Key DDIZAANNODHTRB-UHFFFAOYSA-N
Synonym methyl anisate, methyl p-anisate, methyl p-methoxybenzoate, 4-methoxybenzoic acid methyl ester, p-anisic acid, methyl ester, benzoic acid, 4-methoxy-, methyl ester, methyl 4-anisate, p-methoxybenzoic acid methyl ester, p-anisic acid methyl ester, benzoic acid, p-methoxy-, methyl ester
PubChem CID 8499
ChEBI CHEBI:86903
IUPAC Name methyl 4-methoxybenzoate
SMILES COC1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 49°C
Boiling Point 255°C
Quantity 500 g
Formula Weight 166.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl p-Anisate
EINECSNumber (3)-3054
TSCA Yes