missing translation for 'onlineSavingsMsg'
Learn More

Methyl oxalyl chloride, 97%

Catalog No. p-7046138
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1325806 5 g
AAL1325814 25 g
AAL1325822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1325806 Supplier Thermo Scientific Chemicals Supplier No. L1325806

CAS: 5781-53-3 | C3H3ClO3 | 122.504 g/mol

Methyl oxalyl chloride is used as a synthetic reagent. It serves as a reagent in the synthesis of fused coumarins, substituted isoxazoles and heterocycles. Further, it is used in intramolecular Wittig reactions, and iron-mediated cleavage of C-C bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl oxalyl chloride is used as a synthetic reagent. It serves as a reagent in the synthesis of fused coumarins, substituted isoxazoles and heterocycles. Further, it is used in intramolecular Wittig reactions, and iron-mediated cleavage of C-C bonds.

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible with water, oxidizing agents, alcohols and bases.

Chemical Identifiers

CAS 5781-53-3
Molecular Formula C3H3ClO3
Molecular Weight (g/mol) 122.504
MDL Number MFCD00000705
InChI Key ZXUQEPZWVQIOJE-UHFFFAOYSA-N
Synonym methyl oxalyl chloride, methyl chloroglyoxylate, methyl chlorooxoacetate, chloroglyoxylic acid methyl ester, acetic acid, chlorooxo-, methyl ester, methyl chlorocarbonyl formate, methyloxalylchloride, methyl chloro oxo acetate, methylchlorooxoacetate, methyl 2-chloro-2-oxo-acetate
PubChem CID 79846
IUPAC Name methyl 2-chloro-2-oxoacetate
SMILES COC(=O)C(=O)Cl

Specifications

Density 1.332
Boiling Point 115°C to 118°C
Flash Point 46°C (114°F)
Odor Acrid
Refractive Index 1.419
Quantity 5 g
UN Number UN2920
Beilstein 1071541
Sensitivity Moisture sensitive
Solubility Information Miscible with water.
Formula Weight 122.51
Percent Purity 97%
Chemical Name or Material Methyl oxalyl chloride
Show More Show Less
Hazard Category H226-H302-H314
Hazard Statement GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 227-307-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only