accessibility menu, dialog, popup

UserName

CAS RN 5781-53-3

IUPAC Name: methyl oxalochloridate
Synonyms: methyl oxalochloridate
Molecular Weight (g/mol): 122.5
Molecular Formula: C3H3ClO3
InChi Key: ZXUQEPZWVQIOJE-UHFFFAOYSA-N
SMILES: COC(=O)C(Cl)=O
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)