Learn More
Methyl oxalyl chloride, 97%
CAS: 5781-53-3 | C3H3ClO3 | 122.51 g/mol
$318.48 - $318.48
Chemical Identifiers
| CAS | 5781-53-3 |
|---|---|
| Molecular Formula | C3H3ClO3 |
| Molecular Weight (g/mol) | 122.51 |
| MDL Number | MFCD00000705 |
| InChI Key | ZXUQEPZWVQIOJE-UHFFFAOYSA-N |
| Synonym | methyl oxalyl chloride, methyl chloroglyoxylate, methyl chlorooxoacetate, chloroglyoxylic acid methyl ester, acetic acid, chlorooxo-, methyl ester, methyl chlorocarbonyl formate, methyloxalylchloride, methyl chloro oxo acetate, methylchlorooxoacetate, methyl 2-chloro-2-oxo-acetate |
| PubChem CID | 79846 |
| IUPAC Name | methyl 2-chloro-2-oxoacetate |
| SMILES | COC(=O)C(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC166000250
|
Thermo Scientific Chemicals
166000250 |
25 mL | Glass bottle |
Each for $318.48
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
| 5781-53-3 | |
| 122.51 | |
| ZXUQEPZWVQIOJE-UHFFFAOYSA-N | |
| 79846 | |
| COC(=O)C(=O)Cl |
| C3H3ClO3 | |
| MFCD00000705 | |
| methyl oxalyl chloride, methyl chloroglyoxylate, methyl chlorooxoacetate, chloroglyoxylic acid methyl ester, acetic acid, chlorooxo-, methyl ester, methyl chlorocarbonyl formate, methyloxalylchloride, methyl chloro oxo acetate, methylchlorooxoacetate, methyl 2-chloro-2-oxo-acetate | |
| methyl 2-chloro-2-oxoacetate |
Specifications
| 5781-53-3 | |
| 118°C to 120°C | |
| Authentic | |
| Glass bottle | |
| 1.4179 to 1.4199 | |
| MFCD00000705 | |
| 02, 541 | |
| methyl oxalyl chloride, methyl chloroglyoxylate, methyl chlorooxoacetate, chloroglyoxylic acid methyl ester, acetic acid, chlorooxo-, methyl ester, methyl chlorocarbonyl formate, methyloxalylchloride, methyl chloro oxo acetate, methylchlorooxoacetate, methyl 2-chloro-2-oxo-acetate | |
| ZXUQEPZWVQIOJE-UHFFFAOYSA-N | |
| methyl 2-chloro-2-oxoacetate | |
| 79846 | |
| 97% | |
| Methyl oxalyl chloride |
| 1.3300g/mL | |
| 46°C | |
| 96% min. (GC) | |
| C3H3ClO3 | |
| ClCOCO2CH3 | |
| 25 mL | |
| 1.33 | |
| Solubility in water: decomposes | |
| COC(=O)C(=O)Cl | |
| 122.51 | |
| 122.51 | |
| Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 227-307-4
RUO – Research Use Only