missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange 98.0+%, TCI America™

Catalog No. M048925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M048925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M048925G Supplier TCI America Supplier No. M048925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
MDL Number MFCD00007502
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
Synonym methyl orange, orange iii, gold orange, eniamethyl orange, helianthine, methylorange, c.i. acid orange 52, helianthine b, kca methyl orange, methyloranz czech
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 25 g
UN Number 2811
Formula Weight 327.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Orange
EINECSNumber (5)-4278
RTECSNumber DB6327000
TSCA Yes