missing translation for 'onlineSavingsMsg'
Learn More

Methyl Orange, Sodium Salt Grade, ACS Grade, LabChem™

Catalog No. LC169808
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC169808 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC169808 Supplier LabChem Supplier No. LC169808

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS grade methyl orange sodium salt for laboratory use.

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 300°C
CAS Max % 100
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1000kg/m3
Linear Formula 4-NaOSO2C6H4N:NC6H4-4-N(CH3)2
UN Number UN2811
Density 1,000kg/m3
Color Orange
pH 6.5
Solubility Information Soluble in water
Quantity 25 g
Physical Form Powder
Formula Weight 327.34
Grade ACS
Chemical Name or Material Methyl Orange, Sodium Salt Grade
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature