missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange Indicator, Reag. Ph. Eur., Honeywell Fluka™
Indicator, Reag. Ph. Eur.
$104.86
Chemical Identifiers
| CAS | 547-58-0 |
|---|---|
| Molecular Formula | C14H14N3NaO3S |
| Molecular Weight (g/mol) | 327.334 |
| MDL Number | MFCD00007502 |
| InChI Key | STZCRXQWRGQSJD-UHFFFAOYSA-M |
| Synonym | Helianthin |
| PubChem CID | 23673835 |
| IUPAC Name | sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate |
| SMILES | CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 547-58-0 | |
| 327.334 | |
| STZCRXQWRGQSJD-UHFFFAOYSA-M | |
| 23673835 | |
| CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] |
| C14H14N3NaO3S | |
| MFCD00007502 | |
| Helianthin | |
| sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate |
Specifications
| 547-58-0 | |
| MFCD00007502 | |
| 4732884 | |
| STZCRXQWRGQSJD-UHFFFAOYSA-M | |
| sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate | |
| 23673835 | |
| Complies for sensitivity | |
| Orange | |
| Crystalline Powder |
| C14H14N3NaO3S | |
| UN3143 | |
| Helianthin | |
| CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+] | |
| 327.334 | |
| 327.33g/mol | |
| 3.0 to 4.4 | |
| 3.0 to 4.4 (pink to yellow) | |
| Methyl Orange, Reag. Ph. Eur. |
Safety and Handling
P301 + P310
H301
Danger
DOTInformation : 6.1/PGIII
EINECSNumber : 208-925-3
RTECSNumber : DB6327000