missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Orange, pure, indicator

CAS: 547-58-0 | C14H14N3NaO3S | 327.334 g/mol

Supplier:  Thermo Scientific Chemicals 151425000

 View more versions of this product

Catalog No. AC151425000

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Methyl Orange
547-58-0
MFCD00007502
15, 6178
Solubility in water: soluble in hot water. Other solubilities: practically insoluble in alcohol
Authentic
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
23673835
327.32
Glass bottle
Orange
Fine Crystalline Powder
>300.0°C
C14H14N3NaO3S
16, 331
4-[4-(Dimethylamino)phenylazo]benzenesulfonic acid, sodium salt, Acid Orange 52, C.I. 13025, MO, Helianthise
STZCRXQWRGQSJD-UHFFFAOYSA-M
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]
327.334
461 to 469nm (in 0.01N NaOH)
Pure
from pH 3.2 (pink or red)to pH 4.4 (yellow-orange)
500 g

Chemical Identifiers

547-58-0
327.334
STZCRXQWRGQSJD-UHFFFAOYSA-M
23673835
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]
C14H14N3NaO3S
MFCD00007502
4-[4-(Dimethylamino)phenylazo]benzenesulfonic acid, sodium salt, Acid Orange 52, C.I. 13025, MO, Helianthise
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 208-925-3

RTECSNumber : DB6327000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only