missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Orange

CAS: 547-58-0 | C14H14N3NaO3S | 327.334 g/mol

Supplier:  Thermo Scientific Chemicals A1760422

 View more versions of this product

Catalog No. AAA1760422


Only null left

Description

Description

Methyl Orange is a pH indicator used in titrations for acids. It is also used for histological microscopy. Further, it is used as counter stain to crystal violet in staining pollen tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl Orange is a pH indicator used in titrations for acids. It is also used for histological microscopy. Further, it is used as counter stain to crystal violet in staining pollen tubes.

Solubility
Soluble in ethanol. Partially soluble in hot water. Slightly soluble in cold water and pyrimidine. Insoluble in ether and alcohol.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Methyl Orange
547-58-0
MFCD00007502
4732884
Acid Orange 52; C.I. 13025
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]
327.334
327.34
6.5
>300°C
C14H14N3NaO3S
UN3143
14,6105
STZCRXQWRGQSJD-UHFFFAOYSA-M
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
23673835
Weak, Characteristic
100 g

Chemical Identifiers

547-58-0
327.334
STZCRXQWRGQSJD-UHFFFAOYSA-M
23673835
CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]
C14H14N3NaO3S
MFCD00007502
Acid Orange 52; C.I. 13025
sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DYES, SOLID, TOXIC, N.O.S.

RTECSNumber : DB6327000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only