missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Orange

Catalog No. AAA1760414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1760414 25 g
AAA1760422 100 g
AAA1760436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1760414 Supplier Thermo Scientific Chemicals Supplier No. A1760414

CAS: 547-58-0 | C14H14N3NaO3S | 327.334 g/mol

Methyl Orange is a pH indicator used in titrations for acids. It is also used for histological microscopy. Further, it is used as counter stain to crystal violet in staining pollen tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl Orange is a pH indicator used in titrations for acids. It is also used for histological microscopy. Further, it is used as counter stain to crystal violet in staining pollen tubes.

Solubility
Soluble in ethanol. Partially soluble in hot water. Slightly soluble in cold water and pyrimidine. Insoluble in ether and alcohol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
MDL Number MFCD00007502
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
Synonym Acid Orange 52; C.I. 13025
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point >300°C
UN Number UN3143
Beilstein 4732884
Merck Index 14,6105
Solubility Information Soluble in ethanol. Partially soluble in hot water. Slightly soluble in cold water and pyrimidine. Insoluble in ether and alcohol.
Formula Weight 327.34
Odor Weak, Characteristic
pH 6.5
Quantity 25 g
Chemical Name or Material Methyl Orange
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: DYES, SOLID, TOXIC, N.O.S.
EINECSNumber 208-925-3
RTECSNumber DB6327000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only