missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange (0.1% in Water) [for Titration], TCI America™
Specifications
Specifications
| Color | Red |
| pH | 3.1 to 4.4 |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Formula Weight | 327.33 |
| Grade | Reagent |
| Chemical Name or Material | Methyl Orange (0.1% in Water) [for Titration] |
Safety and Handling
| EINECSNumber | (5)-4278 |
| RTECSNumber | DB6327000 |
| TSCA | Yes |