missing translation for 'onlineSavingsMsg'
Learn More

Methyl octanoate, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019472
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019472 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019472 Supplier Merck Emd Millipore Supplier No. 217195MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 111-11-5
Density 0.877 g/mL (at 20°C)
Flash Point 69.0°C - Closed Cup
Physical Form Neat
Linear Formula CH3(CH2)6 COOCH3
MDL Number MFCD00009551
Boiling Point 193°C to 194°C
Synonym Caprylic acid methyl ester; Methyl caprylate; Octanoic acid methyl ester
Type Analytical Standard
Quantity 5 mL
Formula Weight 158.24
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less