missing translation for 'onlineSavingsMsg'
Learn More

Methyl octacosanoate, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014607
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014607 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014607 Supplier Merck Emd Millipore Supplier No. 74701250MG

Specifications

CAS 55682-92-3
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)26 COOCH3
MDL Number MFCD00042896
Melting Point 67°C to 69°C
Synonym Octacosanoic acid methyl ester
Type Analytical Standard
Quantity 250 mg
Formula Weight 438.77
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less