missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl o-Anisate 99.0+%, TCI America™
$63.38 - $568.54
Chemical Identifiers
| CAS | 606-45-1 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.176 |
| MDL Number | MFCD00008423 |
| InChI Key | PFYHAAAQPNMZHO-UHFFFAOYSA-N |
| Synonym | methyl o-anisate, dimethyl salicylate, o-anisic acid, methyl ester, methyl o-methoxybenzoate, o-methoxy methyl benzoate, o-methoxybenzoic acid methyl ester, methylsalicylate methyl ether, 2-methoxybenzoic acid methyl ester, benzoic acid, 2-methoxy-, methyl ester, 2-methoxybenzoic acid, methyl ester |
| PubChem CID | 61151 |
| IUPAC Name | methyl 2-methoxybenzoate |
| SMILES | COC1=CC=CC=C1C(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A048325ML
|
TCI America
A048325ML |
25 mL |
Each for $63.38
|
|
|||||
|
A0483100ML
|
TCI America
A0483100ML |
100 mL |
Each for $193.86
|
|
|||||
|
A0483500ML
|
TCI America
A0483500ML |
500 mL |
Each for $568.54
|
|
|||||
Chemical Identifiers
| 606-45-1 | |
| 166.176 | |
| PFYHAAAQPNMZHO-UHFFFAOYSA-N | |
| 61151 | |
| COC1=CC=CC=C1C(=O)OC |
| C9H10O3 | |
| MFCD00008423 | |
| methyl o-anisate, dimethyl salicylate, o-anisic acid, methyl ester, methyl o-methoxybenzoate, o-methoxy methyl benzoate, o-methoxybenzoic acid methyl ester, methylsalicylate methyl ether, 2-methoxybenzoic acid methyl ester, benzoic acid, 2-methoxy-, methyl ester, 2-methoxybenzoic acid, methyl ester | |
| methyl 2-methoxybenzoate |
Specifications
| 606-45-1 | |
| 228°C | |
| MFCD00008423 | |
| methyl o-anisate, dimethyl salicylate, o-anisic acid, methyl ester, methyl o-methoxybenzoate, o-methoxy methyl benzoate, o-methoxybenzoic acid methyl ester, methylsalicylate methyl ether, 2-methoxybenzoic acid methyl ester, benzoic acid, 2-methoxy-, methyl ester, 2-methoxybenzoic acid, methyl ester | |
| COC1=CC=CC=C1C(=O)OC | |
| 166.176 | |
| 166.18 | |
| Liquid |
| Yellow | |
| C9H10O3 | |
| 25 mL | |
| PFYHAAAQPNMZHO-UHFFFAOYSA-N | |
| methyl 2-methoxybenzoate | |
| 61151 | |
| ≥99.0% (GC) | |
| Methyl o-Anisate |
Safety and Handling
EINECSNumber : (3)-3055
RTECSNumber : DH3549000
TSCA : Yes