missing translation for 'onlineSavingsMsg'
Learn More

Methyl o-Anisate 99.0+%, TCI America™

Catalog No. A048325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A048325ML 25 mL
A0483100ML 100 mL
A0483500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A048325ML Supplier TCI America Supplier No. A048325ML

Chemical Identifiers

CAS 606-45-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00008423
InChI Key PFYHAAAQPNMZHO-UHFFFAOYSA-N
Synonym methyl o-anisate, dimethyl salicylate, o-anisic acid, methyl ester, methyl o-methoxybenzoate, o-methoxy methyl benzoate, o-methoxybenzoic acid methyl ester, methylsalicylate methyl ether, 2-methoxybenzoic acid methyl ester, benzoic acid, 2-methoxy-, methyl ester, 2-methoxybenzoic acid, methyl ester
PubChem CID 61151
IUPAC Name methyl 2-methoxybenzoate
SMILES COC1=CC=CC=C1C(=O)OC

Specifications

Color Yellow
Boiling Point 228°C
Quantity 25 mL
Formula Weight 166.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl o-Anisate
EINECSNumber (3)-3055
RTECSNumber DH3549000
TSCA Yes