missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Nonadecanoate 98.0+%, TCI America™
$270.29 - $576.00
Chemical Identifiers
| CAS | 1731-94-8 |
|---|---|
| Molecular Formula | C20H40O2 |
| Molecular Weight (g/mol) | 312.538 |
| MDL Number | MFCD00009011 |
| InChI Key | BDXAHSJUDUZLDU-UHFFFAOYSA-N |
| Synonym | methyl nonadecan-1-oate, nonadecanoic acid methyl ester, nonadecanoic acid, methyl ester, unii-67fq8vv2l3, methylnonadecanoate, nonadecanoic acid methyl, nonadecanoic acid methylester, nonadecanoic acid,methyl ester, n-nonadecanoic acid methyl ester, 1n-nonadecanoic acid methyl ester |
| PubChem CID | 15610 |
| IUPAC Name | methyl nonadecanoate |
| SMILES | CCCCCCCCCCCCCCCCCCC(=O)OC |
Chemical Identifiers
| 1731-94-8 | |
| 312.538 | |
| BDXAHSJUDUZLDU-UHFFFAOYSA-N | |
| 15610 | |
| CCCCCCCCCCCCCCCCCCC(=O)OC |
| C20H40O2 | |
| MFCD00009011 | |
| methyl nonadecan-1-oate, nonadecanoic acid methyl ester, nonadecanoic acid, methyl ester, unii-67fq8vv2l3, methylnonadecanoate, nonadecanoic acid methyl, nonadecanoic acid methylester, nonadecanoic acid,methyl ester, n-nonadecanoic acid methyl ester, 1n-nonadecanoic acid methyl ester | |
| methyl nonadecanoate |
Specifications
| 1731-94-8 | |
| White | |
| MFCD00009011 | |
| methyl nonadecan-1-oate, nonadecanoic acid methyl ester, nonadecanoic acid, methyl ester, unii-67fq8vv2l3, methylnonadecanoate, nonadecanoic acid methyl, nonadecanoic acid methylester, nonadecanoic acid,methyl ester, n-nonadecanoic acid methyl ester, 1n-nonadecanoic acid methyl ester | |
| CCCCCCCCCCCCCCCCCCC(=O)OC | |
| 312.538 | |
| 312.54 | |
| Crystalline Powder |
| 40°C | |
| C20H40O2 | |
| 5 g | |
| BDXAHSJUDUZLDU-UHFFFAOYSA-N | |
| methyl nonadecanoate | |
| 15610 | |
| ≥98.0% (GC) | |
| Methyl Nonadecanoate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator