missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Nicotinate 98.0+%, TCI America™
$65.24 - $385.86
Chemical Identifiers
| CAS | 93-60-7 |
|---|---|
| Molecular Formula | C7H7NO2 |
| Molecular Weight (g/mol) | 137.138 |
| MDL Number | MFCD00006388 |
| InChI Key | YNBADRVTZLEFNH-UHFFFAOYSA-N |
| Synonym | methyl nicotinate, nikomet, methylnicotinate, nicotinic acid methyl ester, nicometh, methyl 3-pyridinecarboxylate, methyl-nicotinate, 3-pyridinecarboxylic acid, methyl ester, heat spray, nicotinic acid, methyl ester |
| PubChem CID | 7151 |
| IUPAC Name | methyl pyridine-3-carboxylate |
| SMILES | COC(=O)C1=CN=CC=C1 |
Chemical Identifiers
| 93-60-7 | |
| 137.138 | |
| YNBADRVTZLEFNH-UHFFFAOYSA-N | |
| 7151 | |
| COC(=O)C1=CN=CC=C1 |
| C7H7NO2 | |
| MFCD00006388 | |
| methyl nicotinate, nikomet, methylnicotinate, nicotinic acid methyl ester, nicometh, methyl 3-pyridinecarboxylate, methyl-nicotinate, 3-pyridinecarboxylic acid, methyl ester, heat spray, nicotinic acid, methyl ester | |
| methyl pyridine-3-carboxylate |
Specifications
| 93-60-7 | |
| White-Yellow | |
| MFCD00006388 | |
| methyl nicotinate, nikomet, methylnicotinate, nicotinic acid methyl ester, nicometh, methyl 3-pyridinecarboxylate, methyl-nicotinate, 3-pyridinecarboxylic acid, methyl ester, heat spray, nicotinic acid, methyl ester | |
| COC(=O)C1=CN=CC=C1 | |
| 137.138 | |
| 137.14 | |
| Crystalline Powder |
| 41°C | |
| C7H7NO2 | |
| 25 g | |
| YNBADRVTZLEFNH-UHFFFAOYSA-N | |
| methyl pyridine-3-carboxylate | |
| 7151 | |
| ≥98.0% (GC) | |
| Methyl Nicotinate |
Safety and Handling
EINECSNumber : (5)-0732
RTECSNumber : QT1925000
TSCA : Yes
Recommended Storage : Refrigerator