accessibility menu, dialog, popup

UserName

CH3OON

CAS RN 93-60-7

IUPAC Name: methyl pyridine-3-carboxylate
Synonyms: heat spray methyl nicotinate (substance) methyl nicotinate (product) nikomet methyl-nicotinate nicometh methylnicotinate methyl nicotinate preparation
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: YNBADRVTZLEFNH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CN=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)