missing translation for 'onlineSavingsMsg'
Learn More

Methyl nicotinate, 99%

CAS: 93-60-7 | C7H7NO2 | 137.14 g/mol

$204.71 - $204.71

Chemical Identifiers

CAS 93-60-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00006388
InChI Key YNBADRVTZLEFNH-UHFFFAOYSA-N
Synonym methyl nicotinate, nikomet, methylnicotinate, nicotinic acid methyl ester, nicometh, methyl 3-pyridinecarboxylate, methyl-nicotinate, 3-pyridinecarboxylic acid, methyl ester, heat spray, nicotinic acid, methyl ester
PubChem CID 7151
IUPAC Name methyl pyridine-3-carboxylate
SMILES COC(=O)C1=CN=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC127215000
SDS View Documents
Thermo Scientific Chemicals
127215000
500 g Plastic bottle
Each for $204.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

93-60-7
137.14
YNBADRVTZLEFNH-UHFFFAOYSA-N
7151
COC(=O)C1=CN=CC=C1
C7H7NO2
MFCD00006388
methyl nicotinate, nikomet, methylnicotinate, nicotinic acid methyl ester, nicometh, methyl 3-pyridinecarboxylate, methyl-nicotinate, 3-pyridinecarboxylic acid, methyl ester, heat spray, nicotinic acid, methyl ester
methyl pyridine-3-carboxylate

Specifications

93-60-7
Brown to White
95°C
99%
C7H7NO2
500 g
15, 6173
Solubility in water: negligible. Other solubilities: soluble in alcohol and benzene
COC(=O)C1=CN=CC=C1
137.14
137.14
Crystalline Powder or Crystals
38.0°C to 43.0°C
209.0°C
Authentic
Plastic bottle
MFCD00006388
22, 39
methyl nicotinate, nikomet, methylnicotinate, nicotinic acid methyl ester, nicometh, methyl 3-pyridinecarboxylate, methyl-nicotinate, 3-pyridinecarboxylic acid, methyl ester, heat spray, nicotinic acid, methyl ester
YNBADRVTZLEFNH-UHFFFAOYSA-N
methyl pyridine-3-carboxylate
7151
99%
Methyl nicotinate, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-261-8

RTECSNumber : QT1925000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only