missing translation for 'onlineSavingsMsg'
Learn More

Methyl N-Phenylcarbamate 98.0+%, TCI America™

Catalog No. P101025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P101025G 25 g
P1010500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P101025G Supplier TCI America Supplier No. P101025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2603-10-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00027552
InChI Key IAGUPODHENSJEZ-UHFFFAOYSA-N
Synonym N-Phenylcarbamic Acid Methyl Ester
PubChem CID 17451
IUPAC Name methyl N-phenylcarbamate
SMILES COC(=O)NC1=CC=CC=C1

Specifications

Melting Point 47°C
Boiling Point 126°C
Quantity 25 g
Formula Weight 151.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl N-Phenylcarbamate