missing translation for 'onlineSavingsMsg'
Learn More

Methyl n-Octyl Sulfide 98.0+%, TCI America™

Catalog No. M037925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M037925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M037925ML Supplier TCI America Supplier No. M037925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3698-95-1
Molecular Formula C9H20S
Molecular Weight (g/mol) 160.32
MDL Number MFCD00039990
InChI Key AHCJTMBRROLNHV-UHFFFAOYSA-N
PubChem CID 77289
IUPAC Name 1-(methylsulfanyl)octane
SMILES CCCCCCCCSC

Specifications

Color Yellow
Boiling Point 218°C
Quantity 25 mL
Formula Weight 160.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl n-Octyl Sulfide
EINECSNumber (2)-2447
TSCA No