missing translation for 'onlineSavingsMsg'
Learn More

Methyl myristate, 99%, analytical standard for GC

CAS: 124-10-7 | C15H30O2 | 242.40 g/mol

$72.46 - $176.09

Chemical Identifiers

CAS 124-10-7
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.40
MDL Number MFCD00008983
InChI Key ZAZKJZBWRNNLDS-UHFFFAOYSA-N
Synonym methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci
PubChem CID 31284
IUPAC Name methyl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC460360010
View Documents Promotion Details
N/A 1 g
Each for $72.46
Only null left
 
AC460360050
View Documents Promotion Details
N/A 5 g
Each for $176.09
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

124-10-7
242.40
ZAZKJZBWRNNLDS-UHFFFAOYSA-N
31284
CCCCCCCCCCCCCC(=O)OC
C15H30O2
MFCD00008983
methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci
methyl tetradecanoate

Specifications

124-10-7
0.8630g/mL
>110°C
98.5% min. (GC)
C15H30O2
CH3(CH2)12CO2CH3
0.863
Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol
CCCCCCCCCCCCCC(=O)OC
242.40
242.4
Methyl myristate
18.4°C
323.0°C
Authentic
Glass Bottle
1.4340 to 1.4380
MFCD00008983
methyl myristate, tetradecanoic acid, methyl ester, uniphat a50, metholeneat 2495, myristic acid methyl ester, methyl n-tetradecanoate, myristic acid, methyl ester, methyl myristylate, tetradecanoic acid methyl ester, myristic acid, methyl ester 8ci
ZAZKJZBWRNNLDS-UHFFFAOYSA-N
methyl tetradecanoate
31284
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-680-1

SDS
Documents

Documents

RUO – Research Use Only