missing translation for 'onlineSavingsMsg'
Learn More

Methyl (methylthio)methyl sulfoxide, 97%

CAS: 33577-16-1 | C3H8OS2 | 124.22 g/mol

Supplier:  Thermo Scientific Chemicals L0051009

Catalog No. AAL0051009

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl (methylthio)methyl sulfoxide
1.222
>110°C (230°F)
1.552
10 g
Hygroscopic
OTKFCIVOVKCFHR-UHFFFAOYNA-N
methylsulfanyl(methylsulfinyl)methane
99129
97%
33577-16-1
222°C to 226°C
C3H8OS2
MFCD00002091
906789
methyl methylsulfinylmethyl sulfide, methyl methylsulfinyl methyl sulfide, methane, methylsulfinyl methylthio, methyl methylthio methyl sulfoxide, famso, methanesulfinyl-methylsulfanyl-methane, formaldehyde dimethyl mercaptal s-oxide, methylsulfinyl methylthio methane, methyl methylthiomethyl sulphoxide, formaldehyde methyl mercaptal s-oxide
CSCS(C)=O
124.22
124.22

Chemical Identifiers

33577-16-1
124.22
OTKFCIVOVKCFHR-UHFFFAOYNA-N
99129
CSCS(C)=O
C3H8OS2
MFCD00002091
methyl methylsulfinylmethyl sulfide, methyl methylsulfinyl methyl sulfide, methane, methylsulfinyl methylthio, methyl methylthio methyl sulfoxide, famso, methanesulfinyl-methylsulfanyl-methane, formaldehyde dimethyl mercaptal s-oxide, methylsulfinyl methylthio methane, methyl methylthiomethyl sulphoxide, formaldehyde methyl mercaptal s-oxide
methylsulfanyl(methylsulfinyl)methane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 251-577-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only