missing translation for 'onlineSavingsMsg'
Learn More

Methyl methylsulfonylacetate, 98+%

Catalog No. p-7044747
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0803914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0803914 Supplier Thermo Scientific Chemicals Supplier No. L0803914

CAS: 62020-09-1 | C4H8O4S | 152.16 g/mol

Methyl methylsulfonylacetate is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl methylsulfonylacetate is used as an active pharmaceutical ingredient.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 62020-09-1
Molecular Formula C4H8O4S
Molecular Weight (g/mol) 152.16
MDL Number MFCD00051842
InChI Key RQKDASVRZONLNQ-UHFFFAOYSA-N
Synonym methyl methanesulfonylacetate, methyl 2-methanesulfonylacetate, methyl 2-methylsulfonyl acetate, methyl methylsulfonylacetate, methanesulfonylacetic acid methyl ester, acmc-1b4fd, methyl methylsulfonyl acetate, 2-methylsulfonyl-1-methoxyethanone, methylsulfonylacetic acid methyl ester, methanesulfonyl acetic acid methyl ester
PubChem CID 2759918
SMILES COC(=O)CS(C)(=O)=O

Specifications

Melting Point 59°C to 63°C
Quantity 25 g
Beilstein 2412782
Solubility Information Soluble in methanol. Insoluble in water.
IUPAC Name methyl 2-methylsulfonylacetate
Formula Weight 152.17
Percent Purity ≥98%
Chemical Name or Material Methyl methylsulfonylacetate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only