missing translation for 'onlineSavingsMsg'
Learn More

Methyl Methoxyacetate 97.0+%, TCI America™

Catalog No. p-7132412
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M086325ML 25 mL
M0863500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M086325ML Supplier TCI America Supplier No. M086325ML
Only null left

Chemical Identifiers

CAS 6290-49-9
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00008451
InChI Key QRMHDGWGLNLHMN-UHFFFAOYSA-N
Synonym methyl methoxyacetate, acetic acid, methoxy-, methyl ester, methoxyacetic acid methyl ester, methyl-2-methoxyacetate, methoxyacetic acid, methyl ester, acetic acid, 2-methoxy-, methyl ester, unii-n960nq69lf, methoxy acetic acid methyl ester, methyl metoxyacetate, methyl methoxylacetate
PubChem CID 80507
ChEBI CHEBI:34841
IUPAC Name methyl 2-methoxyacetate
SMILES COCC(=O)OC

Specifications

Color Colorless
Boiling Point 132°C
Quantity 25 mL
UN Number 3272
Formula Weight 104.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Methoxyacetate
EINECSNumber (2)-1349
RTECSNumber AI8910000
TSCA Yes