accessibility menu, dialog, popup

UserName

H3COOOCH3

CAS RN 6290-49-9

IUPAC Name: methyl 2-methoxyacetate
Synonyms: methyl methoxyacetate
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: QRMHDGWGLNLHMN-UHFFFAOYSA-N
SMILES: COCC(=O)OC
Boiling Point 
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)