missing translation for 'onlineSavingsMsg'
Learn More

Methyl Methoxyacetate 97.0+%, TCI America™

Catalog No. m086325ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
M086325ML 25 mL
M0863500ML 500 mL
2 options

Catalog No. M086325ML

Supplier: TCI America M086325ML

Only null left

Chemical Identifiers

CAS 6290-49-9
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00008451
InChI Key QRMHDGWGLNLHMN-UHFFFAOYSA-N
Synonym methyl methoxyacetate, acetic acid, methoxy-, methyl ester, methoxyacetic acid methyl ester, methyl-2-methoxyacetate, methoxyacetic acid, methyl ester, acetic acid, 2-methoxy-, methyl ester, unii-n960nq69lf, methoxy acetic acid methyl ester, methyl metoxyacetate, methyl methoxylacetate
PubChem CID 80507
ChEBI CHEBI:34841
IUPAC Name methyl 2-methoxyacetate
SMILES COCC(=O)OC

Specifications

Color Colorless
Boiling Point 132°C
Quantity 25 mL
UN Number 3272
Formula Weight 104.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Methoxyacetate
EINECSNumber (2)-1349
RTECSNumber AI8910000
TSCA Yes