missing translation for 'onlineSavingsMsg'
Learn More

Methyl methoxyacetate, 99%

CAS: 6290-49-9 | C4H8O3 | 104.11 g/mol

$149.69 - $149.69

Chemical Identifiers

CAS 6290-49-9
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00008451
InChI Key QRMHDGWGLNLHMN-UHFFFAOYSA-N
Synonym methyl methoxyacetate, acetic acid, methoxy-, methyl ester, methoxyacetic acid methyl ester, methyl-2-methoxyacetate, methoxyacetic acid, methyl ester, acetic acid, 2-methoxy-, methyl ester, unii-n960nq69lf, methoxy acetic acid methyl ester, methyl metoxyacetate, methyl methoxylacetate
PubChem CID 80507
ChEBI CHEBI:34841
IUPAC Name methyl 2-methoxyacetate
SMILES COCC(=O)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC165091000
SDS View Documents
Thermo Scientific Chemicals
165091000
100 g Glass bottle
Each for $149.69
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6290-49-9
104.11
QRMHDGWGLNLHMN-UHFFFAOYSA-N
80507
methyl 2-methoxyacetate
C4H8O3
MFCD00008451
methyl methoxyacetate, acetic acid, methoxy-, methyl ester, methoxyacetic acid methyl ester, methyl-2-methoxyacetate, methoxyacetic acid, methyl ester, acetic acid, 2-methoxy-, methyl ester, unii-n960nq69lf, methoxy acetic acid methyl ester, methyl metoxyacetate, methyl methoxylacetate
CHEBI:34841
COCC(=O)OC

Specifications

6290-49-9
1.0500g/mL
35°C
99%
C4H8O3
CH3OCH2CO2CH3
100 g
1.05
Solubility in water: soluble
COCC(=O)OC
104.11
CHEBI:34841
99%
Methyl methoxyacetate, 99%
Colorless to Brown
129°C to 130°C
Authentic
Glass bottle
1.3954 to 1.3974
MFCD00008451
03, 236
methyl methoxyacetate, acetic acid, methoxy-, methyl ester, methoxyacetic acid methyl ester, methyl-2-methoxyacetate, methoxyacetic acid, methyl ester, acetic acid, 2-methoxy-, methyl ester, unii-n960nq69lf, methoxy acetic acid methyl ester, methyl metoxyacetate, methyl methoxylacetate
QRMHDGWGLNLHMN-UHFFFAOYSA-N
methyl 2-methoxyacetate
80507
104.11
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-539-9

RTECSNumber : AI8910000

SDS
Documents

Documents

RUO – Research Use Only