missing translation for 'onlineSavingsMsg'
Learn More

Methyl Methanesulfonylacetate 98.0+%, TCI America™

Catalog No. m13325g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
Catalog No. Quantity
M13325G 5 g
1 options

Catalog No. M13325G

Supplier: TCI America M13325G

Only null left

Chemical Identifiers

CAS 62020-09-1
Molecular Formula C4H8O4S
Molecular Weight (g/mol) 152.16
MDL Number MFCD00051842
InChI Key RQKDASVRZONLNQ-UHFFFAOYSA-N
Synonym methyl methanesulfonylacetate, methyl 2-methanesulfonylacetate, methyl 2-methylsulfonyl acetate, methyl methylsulfonylacetate, methanesulfonylacetic acid methyl ester, acmc-1b4fd, methyl methylsulfonyl acetate, 2-methylsulfonyl-1-methoxyethanone, methylsulfonylacetic acid methyl ester, methanesulfonyl acetic acid methyl ester
PubChem CID 2759918
IUPAC Name methyl 2-methanesulfonylacetate
SMILES COC(=O)CS(C)(=O)=O

Specifications

Melting Point 62°C
Color White-Yellow
Quantity 5 g
Formula Weight 152.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl Methanesulfonylacetate