missing translation for 'onlineSavingsMsg'
Learn More

Methyl Methanesulfonate 98.0+%, TCI America™

Supplier:  TCI America M0369100G

 View more versions of this product

Catalog No. M0369100G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methyl Methanesulfonate
Colorless
C2H6O3S
100 g
methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonate
COS(=O)(=O)C
110.127
CHEBI:25255
≥98.0% (GC)
66-27-3
59°C
MFCD00007557
2810
MBABOKRGFJTBAE-UHFFFAOYSA-N
methyl methanesulfonate
4156
110.13
Liquid

Chemical Identifiers

66-27-3
110.127
MBABOKRGFJTBAE-UHFFFAOYSA-N
4156
methyl methanesulfonate
C2H6O3S
MFCD00007557
methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonate
CHEBI:25255
COS(=O)(=O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : PB2625000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions