missing translation for 'onlineSavingsMsg'
Learn More

Methyl Methanesulfonate 98.0+%, TCI America™

Catalog No. p-7132124
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M036925G 25 g
M0369100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M036925G Supplier TCI America Supplier No. M036925G
Only null left

Chemical Identifiers

CAS 66-27-3
Molecular Formula C2H6O3S
Molecular Weight (g/mol) 110.127
MDL Number MFCD00007557
InChI Key MBABOKRGFJTBAE-UHFFFAOYSA-N
Synonym methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonate
PubChem CID 4156
ChEBI CHEBI:25255
IUPAC Name methyl methanesulfonate
SMILES COS(=O)(=O)C

Specifications

Color Colorless
Boiling Point 59°C
Quantity 25 g
UN Number 2810
Formula Weight 110.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Methanesulfonate
RTECSNumber PB2625000
TSCA Yes