missing translation for 'onlineSavingsMsg'
Learn More

Methyl Methacrylate (stabilized with 6-tert-Butyl-2,4-xylenol) 99.8+%, TCI America™

Catalog No. p-7131981
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M008725ML 25 mL
M0087500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M008725ML Supplier TCI America Supplier No. M008725ML
Only null left

Chemical Identifiers

CAS 80-62-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00008587
InChI Key VVQNEPGJFQJSBK-UHFFFAOYSA-N
Synonym methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
PubChem CID 6658
ChEBI CHEBI:34840
IUPAC Name methyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC

Specifications

Melting Point -48°C
Color Colorless
Boiling Point 100°C
Quantity 25 mL
UN Number 1247
Formula Weight 100.12
Percent Purity ≥99.8% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Methacrylate (stabilized with 6-tert-Butyl-2,4-xylenol)
EINECSNumber (2)-1036
RTECSNumber OZ5075000
TSCA Yes
Recommended Storage Refrigerator