missing translation for 'onlineSavingsMsg'
Learn More

Methyl methacrylate, 99%, stab.

Catalog No. p-7020916
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA13030AE 100 mL
AAA13030AP 500 mL
AAA130300F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA13030AE Supplier Thermo Scientific Chemicals Supplier No. A13030AE

CAS: 80-62-6 | C5H8O2 | 100.117 g/mol

Methyl methacrylate is used as a monomer in the manufacturing of poly(methyl methacrylate) (PMMA). It is also used in the manufacturing of the co-polymer methyl methacrylate-butadiene-styrene (MBS), which is a modifier for polyvinyl chloride. It plays an important role as the cement used in total hip replacements as well as total knee replacements. It is also involved in the preparation of other methacrylates viz. ethyl methacrylate (EMA), butyl methacrylate (BMA) and 2-ethyl hexyl methacrylate (2-EHMA) and methacrylic acid (MAA), which finds application as an intermediate in the manufacture of coating polymers and textile applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl methacrylate is used as a monomer in the manufacturing of poly(methyl methacrylate) (PMMA). It is also used in the manufacturing of the co-polymer methyl methacrylate-butadiene-styrene (MBS), which is a modifier for polyvinyl chloride. It plays an important role as the cement used in total hip replacements as well as total knee replacements. It is also involved in the preparation of other methacrylates viz. ethyl methacrylate (EMA), butyl methacrylate (BMA) and 2-ethyl hexyl methacrylate (2-EHMA) and methacrylic acid (MAA), which finds application as an intermediate in the manufacture of coating polymers and textile applications.

Solubility
Slightly miscible with water.

Notes
Incompatible with oxidizing agents, peroxides, amines, bases, acids, reducing agents and halogens.

Chemical Identifiers

CAS 80-62-6
Molecular Formula C5H8O2
MDL Number MFCD00008587
InChI Key VVQNEPGJFQJSBK-UHFFFAOYSA-N
Synonym methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
PubChem CID 6658
ChEBI CHEBI:34840
IUPAC Name methyl 2-methylprop-2-enoate

Specifications

Melting Point -48°C
Density 0.939
Boiling Point 100°C to 101°C
Flash Point 10°C (50°F)
Refractive Index 1.415
Quantity 100 mL
UN Number UN1247
Beilstein 605459
Merck Index 14,5941
Solubility Information Slightly miscible with water.
SMILES CC(=C)C(=O)OC
Molecular Weight (g/mol) 100.117
Formula Weight 100.12
Percent Purity 99%
Chemical Name or Material Methyl methacrylate, stabilized
Show More Show Less
Hazard Category H225-H315-H317-H319-H335
Hazard Statement GHS H Statement
H225-H315-H317-H335
Highly flammable liquid and vapour.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL METHACRYLATE MONOMER, STABILIZED
EINECSNumber 201-297-1
RTECSNumber OZ5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only