Learn More
Methyl methacrylate, 99%, stab.
Description
Methyl methacrylate is used as a monomer in the manufacturing of poly(methyl methacrylate) (PMMA). It is also used in the manufacturing of the co-polymer methyl methacrylate-butadiene-styrene (MBS), which is a modifier for polyvinyl chloride. It plays an important role as the cement used in total hip replacements as well as total knee replacements. It is also involved in the preparation of other methacrylates viz. ethyl methacrylate (EMA), butyl methacrylate (BMA) and 2-ethyl hexyl methacrylate (2-EHMA) and methacrylic acid (MAA), which finds application as an intermediate in the manufacture of coating polymers and textile applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl methacrylate is used as a monomer in the manufacturing of poly(methyl methacrylate) (PMMA). It is also used in the manufacturing of the co-polymer methyl methacrylate-butadiene-styrene (MBS), which is a modifier for polyvinyl chloride. It plays an important role as the cement used in total hip replacements as well as total knee replacements. It is also involved in the preparation of other methacrylates viz. ethyl methacrylate (EMA), butyl methacrylate (BMA) and 2-ethyl hexyl methacrylate (2-EHMA) and methacrylic acid (MAA), which finds application as an intermediate in the manufacture of coating polymers and textile applications.
Solubility
Slightly miscible with water.
Notes
Incompatible with oxidizing agents, peroxides, amines, bases, acids, reducing agents and halogens.
Specifications
Specifications
| Melting Point | -48°C |
| Density | 0.939 |
| Boiling Point | 100°C to 101°C |
| Flash Point | 10°C (50°F) |
| Refractive Index | 1.415 |
| Quantity | 2500 mL |
| UN Number | UN1247 |
| Beilstein | 605459 |
| Merck Index | 14,5941 |
| Solubility Information | Slightly miscible with water. |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H317-H319-H335 |
| Hazard Statement | GHS H Statement H225-H315-H317-H335 Highly flammable liquid and vapour. Causes skin irritation. May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL METHACRYLATE MONOMER, STABILIZED |
| EINECSNumber | 201-297-1 |
| RTECSNumber | OZ5075000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only