missing translation for 'onlineSavingsMsg'
Learn More

Methyl Methacrylate, 99%, Spectrum™ Chemical

Catalog No. 18611710
Change view
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611710 25 mL Amber Glass Bottle
18611711 500 mL Amber Glass Bottle
18611706 1 L Amber Glass Bottle
18611707 2.5 L Amber Glass Bottle
18611708 20 L Steel Can
18611709 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611710 Supplier Spectrum Chemical Mfg Cor Supplier No. M127425ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 80-62-6

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 mL
Stabilizer Approx. 25 ppm
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.