missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Methacrylate, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor M127425LTGL
Specifications
| 80-62-6 | |
| Amber Glass Bottle | |
| Approx. 25 ppm | |
| Reagent |
| 1 | |
| 2.5 L | |
| 0.99 |
Chemical Identifiers
| 80-62-6 |
CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.