missing translation for 'onlineSavingsMsg'
Learn More

Methyl Linolenate 70.0+%, TCI America™

Catalog No. p-7131795
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L005125ML 25 mL
L0051100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L005125ML Supplier TCI America Supplier No. L005125ML
Only null left

Chemical Identifiers

CAS 301-00-8
Molecular Formula C19H32O2
Molecular Weight (g/mol) 292.46
MDL Number MFCD00135851
InChI Key DVWSXZIHSUZZKJ-YSTUJMKBSA-N
Synonym methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate
PubChem CID 5319706
IUPAC Name methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
SMILES CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC

Specifications

Color Yellow
Boiling Point 347°C
Quantity 25 mL
Formula Weight 292.46
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Linolenate
EINECSNumber (2)-0798
TSCA Yes