missing translation for 'onlineSavingsMsg'
Learn More

Methyl linolenate, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187562
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013853 1 mL
111002185 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013853 Supplier Merck Emd Millipore Supplier No. 622001ML
Only null left

Specifications

CAS 301-00-8
Density 0.895 g/mL (at 25°C)
Flash Point 113.0°C - closed cup
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2 CH=CH)3(CH2)7 COOCH3
MDL Number MFCD00135851
Boiling Point 182°C (3 mmHg)
Synonym Linolenic acid methyl ester; Methyl cis,cis,cis-9,12,15-octadecatrienoate
Type Analytical Standard
Quantity 1 mL
Formula Weight 292.46
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less