missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Linoleate 95.0+%, TCI America™
$178.48 - $1,756.23
Chemical Identifiers
| CAS | 112-63-0 |
|---|---|
| Molecular Formula | C19H34O2 |
| Molecular Weight (g/mol) | 294.479 |
| MDL Number | MFCD00009534 |
| InChI Key | WTTJVINHCBCLGX-NQLNTKRDSA-N |
| Synonym | methyl linoleate, linoleic acid methyl ester, methyl lineoleate, methyl 9z,12z-octadeca-9,12-dienoate, methyl octadecadienoate, methyl 9-cis,12-cis-octadecadienoate, linoleic acid,methyl ester, linoleic acid, methyl ester, methyl linoleate, native, 9,12-octadecadienoic acid z,z-, methyl ester |
| PubChem CID | 5284421 |
| ChEBI | CHEBI:69080 |
| IUPAC Name | methyl (9Z,12Z)-octadeca-9,12-dienoate |
| SMILES | CCCCCC=CCC=CCCCCCCCC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
L007825ML
|
N/A | 25 mL |
Each for $178.48
|
|
|||||
|
L0078500ML
|
N/A | 500 mL |
Each for $1,756.23
|
|
|||||
Chemical Identifiers
| C19H34O2 | |
| MFCD00009534 | |
| methyl linoleate, linoleic acid methyl ester, methyl lineoleate, methyl 9z,12z-octadeca-9,12-dienoate, methyl octadecadienoate, methyl 9-cis,12-cis-octadecadienoate, linoleic acid,methyl ester, linoleic acid, methyl ester, methyl linoleate, native, 9,12-octadecadienoic acid z,z-, methyl ester | |
| CHEBI:69080 | |
| CCCCCC=CCC=CCCCCCCCC(=O)OC |
Specifications
| 112-63-0 | |
| Yellow | |
| C19H34O2 | |
| methyl linoleate, linoleic acid methyl ester, methyl lineoleate, methyl 9z,12z-octadeca-9,12-dienoate, methyl octadecadienoate, methyl 9-cis,12-cis-octadecadienoate, linoleic acid,methyl ester, linoleic acid, methyl ester, methyl linoleate, native, 9,12-octadecadienoic acid z,z-, methyl ester | |
| CCCCCC=CCC=CCCCCCCCC(=O)OC | |
| 294.479 | |
| CHEBI:69080 | |
| ≥95.0% (GC) | |
| Methyl Linoleate |
| -35°C | |
| 346°C | |
| MFCD00009534 | |
| WTTJVINHCBCLGX-NQLNTKRDSA-N | |
| methyl (9Z,12Z)-octadeca-9,12-dienoate | |
| 5284421 | |
| 294.48 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : Yes