missing translation for 'onlineSavingsMsg'
Learn More

Methyl Linoleate 98.0+%, TCI America™

Catalog No. S03251G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03251G 1 g
S03255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03251G Supplier TCI America Supplier No. S03251G

Chemical Identifiers

CAS 112-63-0
Molecular Formula C19H34O2
Molecular Weight (g/mol) 294.479
MDL Number MFCD00009534
InChI Key WTTJVINHCBCLGX-NQLNTKRDSA-N
Synonym methyl linoleate, linoleic acid methyl ester, methyl lineoleate, methyl 9z,12z-octadeca-9,12-dienoate, methyl octadecadienoate, methyl 9-cis,12-cis-octadecadienoate, linoleic acid,methyl ester, linoleic acid, methyl ester, methyl linoleate, native, 9,12-octadecadienoic acid z,z-, methyl ester
PubChem CID 5284421
ChEBI CHEBI:69080
IUPAC Name methyl (9Z,12Z)-octadeca-9,12-dienoate
SMILES CCCCCC=CCC=CCCCCCCCC(=O)OC

Specifications

Melting Point -35°C
Color Yellow
Boiling Point 346°C
Quantity 1 g
Formula Weight 294.48
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Linoleate [Standard Material for GC]
EINECSNumber (2)-0798
TSCA Yes
Recommended Storage Refrigerator