missing translation for 'onlineSavingsMsg'
Learn More

Methyl linoleate, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012502
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012502 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012502 Supplier Merck Emd Millipore Supplier No. 622805ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methyl linoleate, non-conjugated, is more easily oxidized than conjugated methyl linoleate .

Specifications

CAS 112-63-0
Density 0.889 g/mL (at 25°C)
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)3(CH2 CH=CH)2(CH2)7 CO2 CH3
MDL Number MFCD00009534
Melting Point -35°C
Boiling Point 192°C (4 mmHg)
Synonym Linoleic acid methyl ester; Methyl cis,cis-9,12-octadecadienoate
Type Analytical Standard
Quantity 5 mL
Formula Weight 294.47
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less