missing translation for 'onlineSavingsMsg'
Learn More

Methyl linoleate, 95%

CAS: 112-63-0 | C19H34O2 | 294.479 g/mol

Supplier:  Thermo Scientific Chemicals L0745006

 View more versions of this product

Catalog No. AAL0745006


Only null left

Description

Description

The oxidation of methyl linoleate in organic solution and phosphatidylcholine (PC) in liposomal membranes induced by an azo initiator has often been used as a model for lipid peroxidation. Methyl linoleate may be used as a model compound in oxidation/peroxidation assays to evaluate the anti-peroxidation activity of fullerenes, ellagitannins and other natural products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The oxidation of methyl linoleate in organic solution and phosphatidylcholine (PC) in liposomal membranes induced by an azo initiator has often been used as a model for lipid peroxidation. Methyl linoleate may be used as a model compound in oxidation/peroxidation assays to evaluate the anti-peroxidation activity of fullerenes, ellagitannins and other natural products.

Solubility
Soluble in DMF, fat solvents, oils.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

Methyl linoleate
-35°C
207°C to 208°C (11 mmHg)
C19H34O2
MFCD00009534
1727614
methyl linoleate, linoleic acid methyl ester, methyl lineoleate, methyl 9z,12z-octadeca-9,12-dienoate, methyl octadecadienoate, methyl 9-cis,12-cis-octadecadienoate, linoleic acid,methyl ester, linoleic acid, methyl ester, methyl linoleate, native, 9,12-octadecadienoic acid z,z-, methyl ester
WTTJVINHCBCLGX-NQLNTKRDSA-N
methyl (9Z,12Z)-octadeca-9,12-dienoate
5284421
294.47
112-63-0
0.888
200°C (392°F)
1.464
5 g
14,6095
Soluble in DMF,fat solvents,oils.
CCCCCC=CCC=CCCCCCCCC(=O)OC
294.479
CHEBI:69080
95%

Chemical Identifiers

112-63-0
294.479
WTTJVINHCBCLGX-NQLNTKRDSA-N
5284421
methyl (9Z,12Z)-octadeca-9,12-dienoate
C19H34O2
MFCD00009534
methyl linoleate, linoleic acid methyl ester, methyl lineoleate, methyl 9z,12z-octadeca-9,12-dienoate, methyl octadecadienoate, methyl 9-cis,12-cis-octadecadienoate, linoleic acid,methyl ester, linoleic acid, methyl ester, methyl linoleate, native, 9,12-octadecadienoic acid z,z-, methyl ester
CHEBI:69080
CCCCCC=CCC=CCCCCCCCC(=O)OC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-993-0

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only