missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Levulinate 99.0+%, TCI America™
$70.25 - $524.02
Chemical Identifiers
| CAS | 624-45-3 |
|---|---|
| Molecular Formula | C6H10O3 |
| Molecular Weight (g/mol) | 130.143 |
| MDL Number | MFCD00017499 |
| InChI Key | UAGJVSRUFNSIHR-UHFFFAOYSA-N |
| Synonym | methyl levulinate, methyllevulinate, levulinic acid methyl ester, pentanoic acid, 4-oxo-, methyl ester, methyl 4-oxovalerate, levulinic acid, methyl ester, 4-oxovaleric acid methyl ester, 4-oxo-pentanoic acid methyl ester, methyl levulate, me ester |
| PubChem CID | 69354 |
| IUPAC Name | methyl 4-oxopentanoate |
| SMILES | CC(=O)CCC(=O)OC |
Chemical Identifiers
| 624-45-3 | |
| 130.143 | |
| UAGJVSRUFNSIHR-UHFFFAOYSA-N | |
| 69354 | |
| CC(=O)CCC(=O)OC |
| C6H10O3 | |
| MFCD00017499 | |
| methyl levulinate, methyllevulinate, levulinic acid methyl ester, pentanoic acid, 4-oxo-, methyl ester, methyl 4-oxovalerate, levulinic acid, methyl ester, 4-oxovaleric acid methyl ester, 4-oxo-pentanoic acid methyl ester, methyl levulate, me ester | |
| methyl 4-oxopentanoate |
Specifications
| 624-45-3 | |
| 196°C | |
| MFCD00017499 | |
| methyl levulinate, methyllevulinate, levulinic acid methyl ester, pentanoic acid, 4-oxo-, methyl ester, methyl 4-oxovalerate, levulinic acid, methyl ester, 4-oxovaleric acid methyl ester, 4-oxo-pentanoic acid methyl ester, methyl levulate, me ester | |
| CC(=O)CCC(=O)OC | |
| 130.143 | |
| 130.14 | |
| Liquid |
| Yellow | |
| C6H10O3 | |
| 25 g | |
| UAGJVSRUFNSIHR-UHFFFAOYSA-N | |
| methyl 4-oxopentanoate | |
| 69354 | |
| ≥99.0% (GC) | |
| Methyl Levulinate |
Safety and Handling
EINECSNumber : (2)-1492&(2)-1505
TSCA : No