missing translation for 'onlineSavingsMsg'
Learn More

Methyl Levulinate 99.0+%, TCI America™

Catalog No. L004425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L004425G 25 g
L0044100G 100 g
L0044500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L004425G Supplier TCI America Supplier No. L004425G

Chemical Identifiers

CAS 624-45-3
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00017499
InChI Key UAGJVSRUFNSIHR-UHFFFAOYSA-N
Synonym methyl levulinate, methyllevulinate, levulinic acid methyl ester, pentanoic acid, 4-oxo-, methyl ester, methyl 4-oxovalerate, levulinic acid, methyl ester, 4-oxovaleric acid methyl ester, 4-oxo-pentanoic acid methyl ester, methyl levulate, me ester
PubChem CID 69354
IUPAC Name methyl 4-oxopentanoate
SMILES CC(=O)CCC(=O)OC

Specifications

Color Yellow
Boiling Point 196°C
Quantity 25 g
Formula Weight 130.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Levulinate
EINECSNumber (2)-1492&(2)-1505
TSCA No