missing translation for 'onlineSavingsMsg'
Learn More

Methyl Laurate 98.0+%, TCI America™

Catalog No. p-7131772
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L001525ML 25 mL
L0015500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L001525ML Supplier TCI America Supplier No. L001525ML
Only null left

Chemical Identifiers

CAS 111-82-0
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.349
MDL Number MFCD00008966
InChI Key UQDUPQYQJKYHQI-UHFFFAOYSA-N
Synonym methyl laurate, lauric acid methyl ester, dodecanoic acid, methyl ester, methyl laurinate, methyl dodecylate, methyl n-dodecanoate, uniphat a40, metholene 2296, lauric acid, methyl ester, stepan c40
PubChem CID 8139
ChEBI CHEBI:87494
IUPAC Name methyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC

Specifications

Melting Point 5°C
Color Yellow
Boiling Point 262°C
Quantity 25 mL
Formula Weight 214.35
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Laurate
EINECSNumber (2)-0798
RTECSNumber OF0670000
TSCA Yes