missing translation for 'onlineSavingsMsg'
Learn More

Methyl Laurate 99.5+%, TCI America™

Catalog No. S03095ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03095ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03095ML Supplier TCI America Supplier No. S03095ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-82-0
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.349
MDL Number MFCD00008966
InChI Key UQDUPQYQJKYHQI-UHFFFAOYSA-N
Synonym methyl laurate, lauric acid methyl ester, dodecanoic acid, methyl ester, methyl laurinate, methyl dodecylate, methyl n-dodecanoate, uniphat a40, metholene 2296, lauric acid, methyl ester, stepan c40
PubChem CID 8139
ChEBI CHEBI:87494
IUPAC Name methyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC

Specifications

Melting Point 5°C
Color Colorless
Boiling Point 262°C
Quantity 5 mL
Formula Weight 214.35
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Methyl Laurate [Standard Material for GC]
EINECSNumber (2)-0798
RTECSNumber OF0670000
TSCA Yes