missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl L-Pyroglutamate 98.0+%, TCI America™
Supplier: TCI America M2198100G
Specifications
| Methyl L-Pyroglutamate | |
| Yellow | |
| C17H14F3N3S2 | |
| 100 g | |
| JLGLYOGXDCENSE-UHFFFAOYSA-N | |
| N'-methyl-N-[3-(trifluoromethyl)phenyl]{[(4-isothiocyanatophenyl)methyl]sulfanyl}methanimidamide | |
| 78646 | |
| ≥98.0% (GC) |
| 4931-66-2 | |
| 135°C | |
| MFCD00080931 | |
| methyl l-pyroglutamate, methyl s-+-2-pyrrolidone-5-carboxylate, l-pyroglutamic acid methyl ester, methyl pyroglutamate, methyl 5-oxo-l-prolinate, s-methyl 5-oxopyrrolidine-2-carboxylate, proline, 5-oxo-, methyl ester, methyl 2s-5-oxopyrrolidine-2-carboxylate, h-pyr-ome, s-+-methyl 2-pyrrolidone-5-carboxylate | |
| CN=C(NC1=CC=CC(=C1)C(F)(F)F)SCC1=CC=C(C=C1)N=C=S | |
| 381.44 | |
| 143.14 | |
| Liquid |
Chemical Identifiers
| 4931-66-2 | |
| 381.44 | |
| JLGLYOGXDCENSE-UHFFFAOYSA-N | |
| 78646 | |
| CN=C(NC1=CC=CC(=C1)C(F)(F)F)SCC1=CC=C(C=C1)N=C=S |
| C17H14F3N3S2 | |
| MFCD00080931 | |
| methyl l-pyroglutamate, methyl s-+-2-pyrrolidone-5-carboxylate, l-pyroglutamic acid methyl ester, methyl pyroglutamate, methyl 5-oxo-l-prolinate, s-methyl 5-oxopyrrolidine-2-carboxylate, proline, 5-oxo-, methyl ester, methyl 2s-5-oxopyrrolidine-2-carboxylate, h-pyr-ome, s-+-methyl 2-pyrrolidone-5-carboxylate | |
| N'-methyl-N-[3-(trifluoromethyl)phenyl]{[(4-isothiocyanatophenyl)methyl]sulfanyl}methanimidamide |
Safety and Handling
TSCA : No