missing translation for 'onlineSavingsMsg'
Learn More

Methyl L-Pyroglutamate 98.0+%, TCI America™

Catalog No. p-7133086
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M219825G 25 g
M2198100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M219825G Supplier TCI America Supplier No. M219825G
Only null left

Chemical Identifiers

CAS 4931-66-2
Molecular Formula C17H14F3N3S2
Molecular Weight (g/mol) 381.44
MDL Number MFCD00080931
InChI Key JLGLYOGXDCENSE-UHFFFAOYSA-N
Synonym methyl l-pyroglutamate, methyl s-+-2-pyrrolidone-5-carboxylate, l-pyroglutamic acid methyl ester, methyl pyroglutamate, methyl 5-oxo-l-prolinate, s-methyl 5-oxopyrrolidine-2-carboxylate, proline, 5-oxo-, methyl ester, methyl 2s-5-oxopyrrolidine-2-carboxylate, h-pyr-ome, s-+-methyl 2-pyrrolidone-5-carboxylate
PubChem CID 78646
IUPAC Name N'-methyl-N-[3-(trifluoromethyl)phenyl]{[(4-isothiocyanatophenyl)methyl]sulfanyl}methanimidamide
SMILES CN=C(NC1=CC=CC(=C1)C(F)(F)F)SCC1=CC=C(C=C1)N=C=S

Specifications

Color Yellow
Boiling Point 135°C
Quantity 25 g
Formula Weight 143.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl L-Pyroglutamate